Thermo Scientific Chemicals

1-Chloro-6-iodohexane, 97%, Thermo Scientific Chemicals

Catalog number: L05198.09
10 g, Each
Thermo Scientific Chemicals

1-Chloro-6-iodohexane, 97%, Thermo Scientific Chemicals

Catalog number: L05198.09
10 g, Each
Quantity
Catalog number: L05198.09
also known as L05198-09
Price (USD)
Price: 130.00
Online price: 110.65
Your price:
Quantity
-

Chemical Identifiers

CAS
34683-73-3
IUPAC Name
1-chloro-6-iodohexane
Molecular Formula
C6H12ClI
InChI Key
QTJHNJCILMMRIQ-UHFFFAOYSA-N
SMILES
ClCCCCCCI
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5200-1.5265 @ 20?C

Description

1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store at -20°C. Light Sensitive. Store away from oxidizing agents. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text