Thermo Scientific Chemicals

1,9-Nonanediol, 97%, Thermo Scientific Chemicals

Catalog number: L06336.14
25 g, Each
Thermo Scientific Chemicals

1,9-Nonanediol, 97%, Thermo Scientific Chemicals

Catalog number: L06336.14
25 g, Each
Quantity
Catalog number: L06336.14
also known as L06336-14
Price (USD)
Price: 45.60
Online price: 43.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3937-56-2
IUPAC Name
nonane-1,9-diol
Molecular Formula
C9H20O2
InChI Key
ALVZNPYWJMLXKV-UHFFFAOYSA-N
SMILES
OCCCCCCCCCO
Appearance (Color)
White
Form
Crystalline powder or flakes or fused solid or pellets or prills
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Melting Point (clear melt)
45-49?C

Description

1,9-Nonanediol is used in organic synthesis. It also can react with 3,4-dihydro-2H-pyran and 9-(tetrahydropyrane-2-yloxy)nonan-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,9-Nonanediol is used in organic synthesis. It also can react with 3,4-dihydro-2H-pyran and 9-(tetrahydropyrane-2-yloxy)nonan-1-ol.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text