Thermo Scientific Chemicals

N-(2-Hydroxyethyl)hexamethyleneimine, 95%, Thermo Scientific Chemicals

Catalog number: L06497.06
5 g, Each
Thermo Scientific Chemicals

N-(2-Hydroxyethyl)hexamethyleneimine, 95%, Thermo Scientific Chemicals

Catalog number: L06497.06
5 g, Each
Quantity
Catalog number: L06497.06
also known as L06497-06
Price (USD)
Price: 38.10
Online price: 32.65
Your price:
Quantity
-

Chemical Identifiers

CAS
20603-00-3
IUPAC Name
2-(azepan-1-yl)ethan-1-ol
Molecular Formula
C8H17NO
InChI Key
VMRYMOMQCYSPHS-UHFFFAOYSA-N
SMILES
OCCN1CCCCCC1
Form
Liquid or viscous liquid
Identification (FTIR)
Conforms
Assay (GC)
≥94.0%
Refractive Index
1.4820-1.4880 @ 20?C
Appearance (Color)
Clear colorless to yellow

Description

N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(2-Hydroxyethyl)hexamethyleneimine is used in organic synthesis, building blocks. It is mainly involved as an intermediate in the synthesis of Bazedoxifene acetate.

Solubility
Fully miscible in water.

Notes
Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text