Thermo Scientific Chemicals

Cyclohexanemethanol, 99%, Thermo Scientific Chemicals

Catalog number: L06538.22
100 g, Each
Thermo Scientific Chemicals

Cyclohexanemethanol, 99%, Thermo Scientific Chemicals

Catalog number: L06538.22
100 g, Each
Quantity
Catalog number: L06538.22
also known as L06538-22
Price (USD)
Price: 123.00
Online price: 104.65
Your price:
Quantity
-

Chemical Identifiers

CAS
100-49-2
IUPAC Name
cyclohexylmethanol
Molecular Formula
C7H14O
InChI Key
VSSAZBXXNIABDN-UHFFFAOYSA-N
SMILES
OCC1CCCCC1
Form
Liquid or viscous liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%
Refractive Index
1.4625-1.4655 @ 20?C

Description

Cyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.

Solubility
Soluble in Ether, Alcohols.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, Acid chlorides, Acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text