Thermo Scientific Chemicals

1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals

Catalog number: L07033.14
25 g, Each
Thermo Scientific Chemicals

1,2-Epoxy-7-octene, 97%, Thermo Scientific Chemicals

Catalog number: L07033.14
25 g, Each
Quantity
Catalog number: L07033.14
also known as L07033-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
19600-63-6
IUPAC Name
2-(hex-5-en-1-yl)oxirane
Molecular Formula
C8H14O
InChI Key
UKTHULMXFLCNAV-UHFFFAOYNA-N
SMILES
C=CCCCCC1CO1
Appearance (Color)
Clear colorless
Assay (GC)
≥96.0%
Refractive Index
1.4335-1.4385 @ 20?C
Form
Liquid

Description

1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Epoxy-7-octene is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text