Thermo Scientific Chemicals

Glycolaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals

Catalog number: L07099.06
5 g, Each
Thermo Scientific Chemicals

Glycolaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals

Catalog number: L07099.06
5 g, Each
Quantity
Catalog number: L07099.06
also known as L07099-06
Price (USD)
Price: 114.00
Online price: 96.65
Your price:
Quantity
-

Chemical Identifiers

CAS
30934-97-5
IUPAC Name
2,2-dimethoxyethan-1-ol
Molecular Formula
C4H10O3
InChI Key
NYPNCQTUZYWFGG-UHFFFAOYSA-N
SMILES
COC(CO)OC
Form
Liquid
Refractive Index
1.4110-1.4150 @ 20?C
Appearance (Color)
Clear or slightly turbid, colorless to pale yellow
Comment
Product may appear slightly turbid or contain sediment upon storage due to presence of stabiliser.
Assay (GC)
>97.5%

Description

Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycolaldehyde dimethyl acetal is used in the preparation of aceto- xyacetaldehyde dimethyl acetal by reacting with acetic anhydride.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text