Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid
Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid
Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid
Thermo Scientific Chemicals

Chlorendic anhydride, 95%, may cont. up to 3% chlorendic acid

CAS: 115-27-5 | C9H2Cl6O3 | 370.812 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number L07349.0B
also known as L07349-0B
Price (USD)
105.65
Online Exclusive
117.00
Save 11.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
105.65
Online Exclusive
117.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS115-27-5
IUPAC Name1,7,8,9,10,10-hexachloro-4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular FormulaC9H2Cl6O3
InChI KeyFLBJFXNAEMSXGL-UHFFFAOYNA-N
SMILESClC1=C(Cl)C2(Cl)C3C(C(=O)OC3=O)C1(Cl)C2(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder and/or Lumps
Assay (Titration ex Anhydride)>94.0 to <106.0%
Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorendic anhydride is used in the synthesis of polymers, used in production of fiberglass-reinforced resins for chemical industry equipment. It can be used to make alkyd resins for use in special inks and paints. It is used as a hardening agent in epoxy resins used in manufacture of printed circuit boards

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Hans Weiss. Anhydride Curing Agents for Epoxy Resins. Ind. Eng. Chem. 1957, 49, (7), 1089-1090.
  2. P.A. Cusack.; M.S. Heer.; A.W. Monk. Zinc hydroxystannate as an alternative synergist to antimony trioxide in polyester resins containing halogenated flame retardants. Polymer Degradation and Stability. 1997, 58, (1-2), 229-237.