Thermo Scientific Chemicals

1,2-Octanediol, 97%, Thermo Scientific Chemicals

Catalog number: L08031.09
10 g, Each
Thermo Scientific Chemicals

1,2-Octanediol, 97%, Thermo Scientific Chemicals

Catalog number: L08031.09
10 g, Each
Quantity
Catalog number: L08031.09
also known as L08031-09
Price (USD)
Price: 30.20
Online price: 25.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1117-86-8
IUPAC Name
octane-1,2-diol
Molecular Formula
C8H18O2
InChI Key
AEIJTFQOBWATKX-UHFFFAOYNA-N
SMILES
CCCCCCC(O)CO
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White

Description

1,2-Octanediol was used as organic modifier to improve the HPLC separation of organic acids and bases. It was also used in preparation of halohydrin palmitates. It is useful for treating head louse infestation clinically. It is also used in coating materials, slurries, paper mills and water circulation systems for the effective preservation against bacteria and fungi. It is used as an emollient, humectant, and wetting agent in cosmetic and skin care products

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (3 g/L) at 20°C.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text