N-tert-Butylacetamide, 98%
N-tert-Butylacetamide, 98%
N-tert-Butylacetamide, 98%
Thermo Scientific Chemicals

N-tert-Butylacetamide, 98%

CAS: 762-84-5 | C6H13NO | 115.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L08505.09
also known as L08505-09
Price (USD)
80.20
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
80.20
Each
Chemical Identifiers
CAS762-84-5
IUPAC NameN-tert-butylacetamide
Molecular FormulaC6H13NO
InChI KeyACYFWRHALJTSCF-UHFFFAOYSA-N
SMILESCC(=O)NC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)89-100?C
Appearance (Color)White
Assay (GC)≥97.5%
It finds its application in the synthesis of amides using the Ritter reaction with bismuth triflate catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of amides using the Ritter reaction with bismuth triflate catalysis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Branislav Jović, et al. FTIR spectroscopic study of hydrogen bonding and solvent induced frequency shifts of N-tert-butylacetamide.J. Mol. Struct.,2013,1044140-143.
  2. Emmanuel Callens, et al. Synthesis of amides using the Ritter reaction with bismuth triflate catalysis.Tetrahedron Lett.,2006,47(49), 8699-8701.