Thermo Scientific Chemicals

5alpha-Cholestan-3beta-ol, 98%, Thermo Scientific Chemicals

Catalog number: L08624.06
5 g, Each
Thermo Scientific Chemicals

5alpha-Cholestan-3beta-ol, 98%, Thermo Scientific Chemicals

Catalog number: L08624.06
5 g, Each
Quantity
Catalog number: L08624.06
also known as L08624-06
Price (USD)
Price: 79.90
Online price: 67.65
Your price:
Quantity
-

Chemical Identifiers

CAS
80-97-7
IUPAC Name
9a,11a-dimethyl-1-(6-methylheptan-2-yl)-hexadecahydro-1H-cyclopenta[a]phenanthren-7-ol
Molecular Formula
C27H48O
InChI Key
QYIXCDOBOSTCEI-UHFFFAOYNA-N
SMILES
CC(C)CCCC(C)C1CCC2C3CCC4CC(O)CCC4(C)C3CCC12C
Purity (DSC)
≥97.5%
Optical Rotation
24.5 ± 2.5? (C=1 in chloroform)
Form
Crystals or powder or crystalline powder or granules
Appearance (Color)
White to pale cream

Description

5alpha-Cholestan-3beta-ol is used as a standard in lipid analysis using HPLC. It acts as a derivitized steroid compound. It is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5alpha-Cholestan-3beta-ol is used as a standard in lipid analysis using HPLC. It acts as a derivitized steroid compound. It is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text