Thermo Scientific Chemicals

N-Boc-D-leucine hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: L09124.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-D-leucine hydrate, 98+%, Thermo Scientific Chemicals

Catalog number: L09124.06
5 g, Each
Quantity
Catalog number: L09124.06
also known as L09124-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
16937-99-8
IUPAC Name
2-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid hydrate
Molecular Formula
C11H23NO5
InChI Key
URQQEIOTRWJXBA-UHFFFAOYNA-N
SMILES
O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
3.75-10.46% (0.5-1.5 waters)
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Optical Rotation
25.2 ± 0.1? (C=2 in acetic acid)
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text