1,1,3,3-Tetraethylurea, 99%
1,1,3,3-Tetraethylurea, 99%
1,1,3,3-Tetraethylurea, 99%
Thermo Scientific Chemicals

1,1,3,3-Tetraethylurea, 99%

CAS: 1187-03-7 | C9H20N2O | 172.272 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09458.14
also known as L09458-14
Price (USD)
103.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
103.00
Each
Chemical Identifiers
CAS1187-03-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Identification (FTIR)Conforms
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.4460-1.4490 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only