Thermo Scientific Chemicals

Bromomethyl methyl ether, tech. 90%, Thermo Scientific Chemicals

Catalog number: L09837.14
25 g, Each
Thermo Scientific Chemicals

Bromomethyl methyl ether, tech. 90%, Thermo Scientific Chemicals

Catalog number: L09837.14
25 g, Each
Quantity
Catalog number: L09837.14
also known as L09837-14
Price (USD)
Price: 158.00
Online price: 135.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13057-17-5
IUPAC Name
bromo(methoxy)methane
Molecular Formula
C2H5BrO
InChI Key
JAMFGQBENKSWOF-UHFFFAOYSA-N
SMILES
COCBr
Form
Liquid
Assay (GC)
≥88.0%
Appearance (Color)
Clear colorless to yellow

Description

Bromomethyl methyl ether is used as reagent for protection of OH groups as their methoxymethyl (MOM) ethers. It is used in enantiocontrolled synthesis of intermediate containing carbons 18 to 35 of the macrocyclic immunosuppressant FK-506. It was used in the synthesis of 1-methoxymethyl-4-tritylthio-2-azetidinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromomethyl methyl ether is used as reagent for protection of OH groups as their methoxymethyl (MOM) ethers. It is used in enantiocontrolled synthesis of intermediate containing carbons 18 to 35 of the macrocyclic immunosuppressant FK-506. It was used in the synthesis of 1-methoxymethyl-4-tritylthio-2-azetidinone.

Solubility
Sparingly soluble in water.

Notes
Moisture Sensitive. Store at 4°C. Do not store together with acids. Store away from water and strong bases. Incompatible with acids, bases, moisture, heat and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text