Thermo Scientific Chemicals

1,4-Dibromopentane, 97%, Thermo Scientific Chemicals

Catalog number: L09842.14
25 g, Each
Thermo Scientific Chemicals

1,4-Dibromopentane, 97%, Thermo Scientific Chemicals

Catalog number: L09842.14
25 g, Each
Quantity
Catalog number: L09842.14
also known as L09842-14
Price (USD)
Price: 97.30
Online price: 92.65
Your price:
Quantity
-

Chemical Identifiers

CAS
626-87-9
IUPAC Name
1,4-dibromopentane
Molecular Formula
C5H10Br2
InChI Key
CNBFRBXEGGRSPL-UHFFFAOYNA-N
SMILES
CC(Br)CCCBr
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.5065-1.5115 @ 20?C
Identification (FTIR)
Conforms
Form
Liquid

Description

1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromopentane is used in the preparation of pharaoh ant trail pheromone stereoisomer, indolizidine alkaloid. It is also involved in the preparation 1,2-dimethyl-1-phenylcyclopentane and boc-L-2-amino-6-bromoheptanoic acid.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text