Thermo Scientific Chemicals

Bis(2-ethylhexyl) adipate, 99%, Thermo Scientific Chemicals

Catalog number: L10195.22
100 g, Each
Thermo Scientific Chemicals

Bis(2-ethylhexyl) adipate, 99%, Thermo Scientific Chemicals

Catalog number: L10195.22
100 g, Each
Quantity
Catalog number: L10195.22
also known as L10195-22
Price (USD)
Price: 134.00
Online price: 117.65
Your price:
Quantity
-

Chemical Identifiers

CAS
103-23-1
IUPAC Name
1,6-bis(2-ethylhexyl) hexanedioate
Molecular Formula
C22H42O4
InChI Key
SAOKZLXYCUGLFA-UHFFFAOYNA-N
SMILES
CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC
Refractive Index
1.4440-1.4500 @ 20?C
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

It is employed as a plasticizer. It is a skin conditioning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a plasticizer. It is a skin conditioning solvent.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text