Thermo Scientific Chemicals

1,1-Dibromo-3,3,3-trifluoroacetone, 95%, Thermo Scientific Chemicals

Catalog number: L10826.14
25 g, Each
Thermo Scientific Chemicals

1,1-Dibromo-3,3,3-trifluoroacetone, 95%, Thermo Scientific Chemicals

Catalog number: L10826.14
25 g, Each
Quantity
Catalog number: L10826.14
also known as L10826-14
Price (USD)
Price: 326.00
Online price: 281.65
Your price:
Quantity
-

Chemical Identifiers

CAS
431-67-4
IUPAC Name
3,3-dibromo-1,1,1-trifluoropropan-2-one
Molecular Formula
C3HBr2F3O
InChI Key
HEPPAPZASXFWTB-UHFFFAOYSA-N
SMILES
FC(F)(F)C(=O)C(Br)Br
Appearance (Color)
Clear colorless to yellow to orange
Form
Liquid
Assay (GC)
≥94.0%
Refractive Index
1.4275-1.4335 @ 20?C

Description

1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1-Dibromo-3,3,3-trifluoroacetone readily forms trifluoropyruvaldehyde hydrate in the presence of aqueous NaOAc, which is a useful intermediate, e.g. in the synthesis of a fused pyrazine derivative.

Solubility
Soluble in Chloroform. Not miscible or difficult to mix in water.

Notes
Keep container tightly closed. Incompatible with strong bases, active metals and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text