Thermo Scientific Chemicals

2-Fluoro-1-methylpyridinium p-toluenesulfonate, tech. 90%, Thermo Scientific Chemicals

Catalog number: L11088.03
1 g, Each
Thermo Scientific Chemicals

2-Fluoro-1-methylpyridinium p-toluenesulfonate, tech. 90%, Thermo Scientific Chemicals

Catalog number: L11088.03
1 g, Each
Quantity
Catalog number: L11088.03
also known as L11088-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
58086-67-2
IUPAC Name
2-fluoro-1-methylpyridin-1-ium 4-methylbenzene-1-sulfonate
Molecular Formula
C13H14FNO3S
InChI Key
HQWDKLAIDBOLFE-UHFFFAOYSA-M
SMILES
C[N+]1=CC=CC=C1F.CC1=CC=C(C=C1)S([O-])(=O)=O
Appearance (Color)
White
Assay from Supplier's CofA
≥88.0% (US-sourced material)
Elemental Analysis
C: 48.50-60.63% (Theory: 55.11%, UK-sourced material)
Elemental Analysis
N: 4.35-5.44% (Theory: 4.94%, UK-sourced material)
Assay (special)
≥88.0% (based on elemental C, UK-sourced material)

Description

2-Fluoro-1-methylpyridinium p-toluenesulfonate Is a peptide coupling reagent. Also used in the synthesis of ? -ribonucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-1-methylpyridinium p-toluenesulfonate Is a peptide coupling reagent. Also used in the synthesis of ɑ -ribonucleotides.

Solubility
Soluble in water (hydrolyzes), DMSO, dichloromethane, and acetonitrile.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text