Thermo Scientific Chemicals

4-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L11486.14
25 g, Each
Thermo Scientific Chemicals

4-Bromobenzyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L11486.14
25 g, Each
Quantity
Catalog number: L11486.14
also known as L11486-14
Price (USD)
Price: 177.00
Online price: 153.65
Your price:
Quantity
-

Chemical Identifiers

CAS
589-17-3
IUPAC Name
1-bromo-4-(chloromethyl)benzene
Molecular Formula
C7H6BrCl
InChI Key
BSIIGUGKOPPTPZ-UHFFFAOYSA-N
SMILES
ClCC1=CC=C(Br)C=C1
Identification (FTIR)
Conforms
Melting Point (clear melt)
34.0-44.0?C
Appearance (Color)
White to gray
Form
Crystals or powder or crystalline powder or fused solid or flakes
Assay (GC)
≥96.0%

Description

4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text