Thermo Scientific Chemicals

N,N'-Ethylenebisacrylamide, 96%, Thermo Scientific Chemicals

Catalog number: L12544.03
1 g, Each
Thermo Scientific Chemicals

N,N'-Ethylenebisacrylamide, 96%, Thermo Scientific Chemicals

Catalog number: L12544.03
1 g, Each
Quantity
Catalog number: L12544.03
also known as L12544-03
Price (USD)
Price: 75.40
Online price: 64.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2956-58-3
IUPAC Name
N-[2-(prop-2-enamido)ethyl]prop-2-enamide
Molecular Formula
C8H12N2O2
InChI Key
AYGYHGXUJBFUJU-UHFFFAOYSA-N
SMILES
C=CC(=O)NCCNC(=O)C=C
Form
Crystals or powder or crystalline powder
Polymer Test
10% in Methanol: Clear Solution
Assay (HPLC)
≥95.0%
Appearance (Color)
White to pale cream

Description

N,N'-Ethylenebisacrylamide is used in the preparation of N,N'-dicinnamoyl-ethylenediamine using poly(N-octadecylacrylamide) as a catalyst. It is also used as an intermediate in pharmaceutical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Ethylenebisacrylamide is used in the preparation of N,N′-dicinnamoyl-ethylenediamine using poly(N-octadecylacrylamide) as a catalyst. It is also used as an intermediate in pharmaceutical industries.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text