Thermo Scientific Chemicals

1,2-Ethanedithiol, 98+%, Thermo Scientific Chemicals

Catalog number: L12865.36
500 g, Each
Thermo Scientific Chemicals

1,2-Ethanedithiol, 98+%, Thermo Scientific Chemicals

Catalog number: L12865.36
500 g, Each
Quantity
Catalog number: L12865.36
also known as L12865-36
Price (USD)
Price: 327.00
Online price: 277.65
Your price:
Quantity
-

Chemical Identifiers

CAS
540-63-6
IUPAC Name
ethane-1,2-dithiol
Molecular Formula
C2H6S2
InChI Key
VYMPLPIFKRHAAC-UHFFFAOYSA-N
SMILES
SCCS
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow or pale green
Refractive Index
1.5560-1.5600 @ 20?C
Assay (GC)
≥98.0%
Form
Liquid

Description

1,2-Ethanedithiol is used as a reagent in organic synthesis to convert carbonyl compounds to thioacetals (1,3-dithiolanes). It replaces the toxic reagent arsenic tirchloride, which is involved in the synthesis of membrane-permeant fluorogenic biarsenicals from precursor dyes fluorescein and resorufin. It acts bidendate ligand to form complexes with metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Ethanedithiol is used as a reagent in organic synthesis to convert carbonyl compounds to thioacetals (1,3-dithiolanes). It replaces the toxic reagent arsenic tirchloride, which is involved in the synthesis of membrane-permeant fluorogenic biarsenicals from precursor dyes fluorescein and resorufin. It acts bidendate ligand to form complexes with metal ions.

Solubility
Miscible with ethanol, acetone, ether and benzene. Slightly miscible with water.

Notes
Air sensitive. Keep away from sources of ignition. Incompatible with alkali metals. Avoid inhalation of its vapor.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text