Thermo Scientific Chemicals

1-Boc-piperazine, 99%, Thermo Scientific Chemicals

Catalog number: L13363.06
5 g, Each
Thermo Scientific Chemicals

1-Boc-piperazine, 99%, Thermo Scientific Chemicals

Catalog number: L13363.06
5 g, Each
Quantity
Catalog number: L13363.06
also known as L13363-06
Price (USD)
Price: 110.00
Online price: 93.65
Your price:
Quantity
-

Chemical Identifiers

CAS
57260-71-6
Form
Crystals or crystalline powder or lumpy powder or fused solid
Assay (GC)
≥98.5%
Appearance (Color)
White to cream to yellow
Solution Test
5% in ethanol: clear & colourless to yellow

Description

1-Boc-piperazine is useful for the preparation of (m-phenoxy)phenyl substituted piperazine derivatives. It is also used in the synthesis of indazole DNA gyrase inhibitors. Further, it plays an important role in the preparation of alfa,beta-poly(2-oxazoline) lipopolymers through living cationic ring opening polymerization. It is also in involved in Buchwald-Hartwig coupling reactions with aryl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-piperazine is useful for the preparation of (m-phenoxy)phenyl substituted piperazine derivatives. It is also used in the synthesis of indazole DNA gyrase inhibitors. Further, it plays an important role in the preparation of alfa,beta-poly(2-oxazoline) lipopolymers through living cationic ring opening polymerization. It is also in involved in Buchwald-Hartwig coupling reactions with aryl halides.

Solubility
Soluble in ethyl acetate, methanol and water.

Notes
Hygroscopic. Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text