Thermo Scientific Chemicals

4-Aminophthalonitrile, 98%, Thermo Scientific Chemicals

Catalog number: L13433.03
1 g, Each
Thermo Scientific Chemicals

4-Aminophthalonitrile, 98%, Thermo Scientific Chemicals

Catalog number: L13433.03
1 g, Each
Quantity
Catalog number: L13433.03
also known as L13433-03
Price (USD)
Price: 52.60
Online price: 45.65
Your price:
Quantity
-

Chemical Identifiers

CAS
56765-79-8
IUPAC Name
4-aminobenzene-1,2-dicarbonitrile
Molecular Formula
C8H5N3
InChI Key
RRCAJFYQXKPXOJ-UHFFFAOYSA-N
SMILES
NC1=CC=C(C#N)C(=C1)C#N
Appearance (Color)
White to yellow to orange or yellow-brown
Melting Point (clear melt)
178.0-184.0°C
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text