Thermo Scientific Chemicals

1,4-Phenylenediacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L13464.14
25 g, Each
Thermo Scientific Chemicals

1,4-Phenylenediacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L13464.14
25 g, Each
Quantity
Catalog number: L13464.14
also known as L13464-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
622-75-3
IUPAC Name
2-[4-(cyanomethyl)phenyl]acetonitrile
Molecular Formula
C10H8N2
InChI Key
FUQCKESKNZBNOG-UHFFFAOYSA-N
SMILES
N#CCC1=CC=C(CC#N)C=C1
Appearance (Color)
White to cream to yellow to brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
91.0-100.0?C

Description

1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Phenylenediacetonitrile, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text