Thermo Scientific Chemicals

2-Fluoro-1,4-dimethoxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: L13505.06
5 g, Each
Thermo Scientific Chemicals

2-Fluoro-1,4-dimethoxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: L13505.06
5 g, Each
Quantity
Catalog number: L13505.06
also known as L13505-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
82830-49-7
IUPAC Name
2-fluoro-1,4-dimethoxybenzene
Molecular Formula
C8H9FO2
InChI Key
WNCYZVMZKSOPMU-UHFFFAOYSA-N
SMILES
COC1=CC=C(OC)C(F)=C1
Appearance (Color)
Colorless to white to yellow
Form
Crystals or powder or crystalline powder or clear liquid as melt
Identification (FTIR)
Conforms
Refractive Index
1.5020-1.5070 @ 20?C
Assay (GC)
≥96.0%

Description

2-Fluoro-1,4-dimethoxybenzene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-1,4-dimethoxybenzene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text