Thermo Scientific Chemicals

Diethyl sulfate, 98%, Thermo Scientific Chemicals

Catalog number: L14291.AE
100 mL, Each
Thermo Scientific Chemicals

Diethyl sulfate, 98%, Thermo Scientific Chemicals

Catalog number: L14291.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L14291.AE
also known as L14291-AE
Price (USD)
Price: 29.60
Online price: 25.65
Your price:
Quantity
-

Chemical Identifiers

CAS
64-67-5
IUPAC Name
diethyl sulfate
Molecular Formula
C4H10O4S
InChI Key
DENRZWYUOJLTMF-UHFFFAOYSA-N
SMILES
CCOS(=O)(=O)OCC
Refractive Index
1.3985-1.4025 @ 20?C
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Form
Liquid
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text