Thermo Scientific Chemicals

Methyl indole-3-carboxylate, 99%, Thermo Scientific Chemicals

Catalog number: L14516.22
100 g, Each
Thermo Scientific Chemicals

Methyl indole-3-carboxylate, 99%, Thermo Scientific Chemicals

Catalog number: L14516.22
100 g, Each
Quantity
Catalog number: L14516.22
also known as L14516-22
Price (USD)
Price: 176.00
Online price: 150.65
Your price:
Quantity
-

Chemical Identifiers

CAS
942-24-5
IUPAC Name
methyl 1H-indole-3-carboxylate
Molecular Formula
C10H9NO2
InChI Key
QXAUTQFAWKKNLM-UHFFFAOYSA-N
SMILES
COC(=O)C1=CNC2=CC=CC=C12
Form
Powder or lumps
Melting Point (clear melt)
145.0-152.0?C
Assay (HPLC)
≥98.5%
Appearance (Color)
White to cream to dark cream to brown or purple to dark pink to pale pink

Description

Reactant used for preparation of nitric oxide synthesis (nNOS) inhibitors, protein kinase c alpha (PKCα) inhibitors, inhibitors of the C-terminal domain of RNA polymerase II as antitumor agents, kinase insert domain receptor (KDR) inhibitors, organocatalysts for the anti-Mannich reaction, very late antigen-4 (VLA-4) antagonists, inhibitors of Human 5-Lipoxygenase, serotonin 5-HT4 receptor antagonists and hyaluronidase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of nitric oxide synthesis (nNOS) inhibitors, protein kinase c alpha (PKCα) inhibitors, inhibitors of the C-terminal domain of RNA polymerase II as antitumor agents, kinase insert domain receptor (KDR) inhibitors, organocatalysts for the anti-Mannich reaction, very late antigen-4 (VLA-4) antagonists, inhibitors of Human 5-Lipoxygenase, serotonin 5-HT4 receptor antagonists and hyaluronidase inhibitors.

Solubility
Slightly soluble methanol and dimethyl sulfoxide. Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text