Thermo Scientific Chemicals

Sodium vinylsulfonate, tech. ca 25% aq. soln., Thermo Scientific Chemicals

Catalog number: L15356.AU
1000 mL, Each
Thermo Scientific Chemicals

Sodium vinylsulfonate, tech. ca 25% aq. soln., Thermo Scientific Chemicals

Catalog number: L15356.AU
1000 mL, Each
Quantity
Catalog number: L15356.AU
also known as L15356-AU
Price (USD)
Price: 193.00
Online price: 164.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3039-83-6
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Color (APHA)
≤400 APHA
Refractive Index
1.3705-1.3775 @ 20°C
Assay from Suppliers CofA
25-26% active content

Description

Sodium vinylsulfonate is a useful reagent (monomer) for the formation of poly(anionic) polymers and copolymers. It is employed as a basic brightener and leveling agent in nickel baths. It is also used as intermediate for organic synthesis, surfactant, pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium vinylsulfonate is a useful reagent (monomer) for the formation of poly(anionic) polymers and copolymers. It is employed as a basic brightener and leveling agent in nickel baths. It is also used as intermediate for organic synthesis, surfactant, pharmaceutical industry.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text