Thermo Scientific Chemicals

3-Methoxy-3-methyl-1-butanol, 98+%, Thermo Scientific Chemicals

Catalog number: L15953.0F
2500 mL, Each
Thermo Scientific Chemicals

3-Methoxy-3-methyl-1-butanol, 98+%, Thermo Scientific Chemicals

Catalog number: L15953.0F
2500 mL, Each
Quantity
Catalog number: L15953.0F
also known as L15953-0F
Price (USD)
Price: 277.00
Online price: 235.65
Your price:
Quantity
-

Chemical Identifiers

CAS
56539-66-3
IUPAC Name
3-methoxy-3-methylbutan-1-ol
Molecular Formula
C6H14O2
InChI Key
MFKRHJVUCZRDTF-UHFFFAOYSA-N
SMILES
COC(C)(C)CCO
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.4250-1.4290 @ 20?C
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms

Description

3-Methoxy-3-methyl-1-butanol is a solvent that compares favorably with its isomer ethylene glycol monobutyl ether. It is employed as raw material for the production of industrial detergents. It is also used as solvent for paints, inks, and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-3-methyl-1-butanol is a solvent that compares favorably with its isomer ethylene glycol monobutyl ether. It is employed as raw material for the production of industrial detergents. It is also used as solvent for paints, inks, and fragrances.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text