Thermo Scientific Chemicals

1-Benzyloxy-4-iodobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: L16092.06
5 g, Each
Thermo Scientific Chemicals

1-Benzyloxy-4-iodobenzene, 98+%, Thermo Scientific Chemicals

Catalog number: L16092.06
5 g, Each
Quantity
Catalog number: L16092.06
also known as L16092-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
19578-68-8
IUPAC Name
1-(benzyloxy)-4-iodobenzene
Molecular Formula
C13H11IO
InChI Key
MPWFGAWFTAZWKZ-UHFFFAOYSA-N
SMILES
IC1=CC=C(OCC2=CC=CC=C2)C=C1
Melting Point
55.0-60.0°C
Appearance (Color)
Pale amber
Assay from Supplier's CofA
≥98.0%
Form
Solid

Description

1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text