Thermo Scientific Chemicals

N-(4-Chloro-2-butynyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: L16313.06
5 g, Each
Thermo Scientific Chemicals

N-(4-Chloro-2-butynyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog number: L16313.06
5 g, Each
Quantity
Catalog number: L16313.06
also known as L16313-06
Price (USD)
Price: 324.00
Online price: 279.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4819-69-6
IUPAC Name
2-(4-chlorobut-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C12H8ClNO2
InChI Key
TXNDRPKNOXQAAO-UHFFFAOYSA-N
SMILES
ClCC#CCN1C(=O)C2=CC=CC=C2C1=O
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
115.0-121.0?C
Assay (HPLC)
≥96.0%

Description

N-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(4-Chloro-2-butynyl)phthalimide is used to make other chemicals and as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text