Thermo Scientific Chemicals

(S)-(-)-1-(4-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 98%, Thermo Scientific Chemicals

Catalog number: L16322.06
5 g, Each
Thermo Scientific Chemicals

(S)-(-)-1-(4-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 98%, Thermo Scientific Chemicals

Catalog number: L16322.06
5 g, Each
Quantity
Catalog number: L16322.06
also known as L16322-06
Price (USD)
Price: 85.70
Online price: 74.65
Your price:
Quantity
-

Chemical Identifiers

CAS
41851-59-6
IUPAC Name
1-(4-methoxyphenyl)ethan-1-amine
Molecular Formula
C9H13NO
InChI Key
JTDGKQNNPKXKII-UHFFFAOYNA-N
SMILES
COC1=CC=C(C=C1)C(C)N
Appearance (Color)
Clear, colourless
Form
liquid
Optical Rotation
-34? ? 1? (neat)
Assay (GC)
>98.5%
Enantiomeric excess
> 97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text