Thermo Scientific Chemicals

(S)-(-)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%, Thermo Scientific Chemicals

Catalog number: L16324.03
1 g, Each
Thermo Scientific Chemicals

(S)-(-)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%, Thermo Scientific Chemicals

Catalog number: L16324.03
1 g, Each
Quantity
Catalog number: L16324.03
also known as L16324-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
82796-69-8
IUPAC Name
1-(3-methoxyphenyl)ethan-1-amine
Molecular Formula
C9H13NO
InChI Key
CJWGCBRQAHCVHW-UHFFFAOYNA-N
SMILES
COC1=CC=CC(=C1)C(C)N
Optical Rotation
-27° ± 2°(neat)
Appearance (Color)
Clear colourless to very pale yellow
Enantiomeric excess
>99.0% by GC
Assay (GC)
≥99.0%
Refractive Index
1.5325-1.5345 @ 20°C

Description

Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of the (R)-atropisomer, rivastigmine tartrate.

Solubility
Soluble in water (10g/L).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text