Thermo Scientific Chemicals

2-Cyano-4-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: L17148.06
5 g, Each
Thermo Scientific Chemicals

2-Cyano-4-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog number: L17148.06
5 g, Each
Quantity
Catalog number: L17148.06
also known as L17148-06
Price (USD)
Price: 213.00
Your price:
Quantity
-

Chemical Identifiers

CAS
1620-76-4
IUPAC Name
4-methylpyridine-2-carbonitrile
Molecular Formula
C7H6N2
InChI Key
LQAWSWUFSHYCHP-UHFFFAOYSA-N
SMILES
CC1=CC(=NC=C1)C#N
Assay (GC)
≥97.5%
Appearance (Color)
Cream to yellow to pale brown
Form
Crystals or powder or crystalline powder or lumps

Description

2-Cyano-4-methylpyridine can react to produce C7H7NO2.ClH. This reaction will need reagent 6 NHCl.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyano-4-methylpyridine can react to produce C7H7NO2.ClH. This reaction will need reagent 6 NHCl.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text