Thermo Scientific Chemicals

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 98%, Thermo Scientific Chemicals

Catalog number: L17278.06
5 g, Each
Thermo Scientific Chemicals

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 98%, Thermo Scientific Chemicals

Catalog number: L17278.06
5 g, Each
Quantity
Catalog number: L17278.06
also known as L17278-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61676-62-8
IUPAC Name
4,4,5,5-tetramethyl-2-(propan-2-yloxy)-1,3,2-dioxaborolane
Molecular Formula
C9H19BO3
InChI Key
MRWWWZLJWNIEEJ-UHFFFAOYSA-N
SMILES
CC(C)OB1OC(C)(C)C(C)(C)O1
Form
Liquid
Refractive Index
1.4070-1.4110 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥92.0%
Identification (FTIR)
Conforms

Description

Reagent for fluorenylborolane preparation, reagent used to borylate arenes. Reagent used to borylate arenes and for fluorenylborolane preparation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for fluorenylborolane preparation, reagent used to borylate arenes. Reagent used to borylate arenes and for fluorenylborolane preparation.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text