Thermo Scientific Chemicals

5-Iodo-1,2,3-trimethoxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: L17414.03
1 g, Each
Thermo Scientific Chemicals

5-Iodo-1,2,3-trimethoxybenzene, 97%, Thermo Scientific Chemicals

Catalog number: L17414.03
1 g, Each
Quantity
Catalog number: L17414.03
also known as L17414-03
Price (USD)
Price: 84.90
Online price: 72.65
Your price:
Quantity
-

Chemical Identifiers

CAS
25245-29-8
IUPAC Name
5-iodo-1,2,3-trimethoxybenzene
Molecular Formula
C9H11IO3
InChI Key
IWPMQXOVTMABLF-UHFFFAOYSA-N
SMILES
COC1=CC(I)=CC(OC)=C1OC
Appearance (Color)
Yellow to pale brown
Form
Crystals or powder or crystalline powder or granules
Assay (GC)
≥96.0%
Melting Point (clear melt)
80.5-89.5?C

Description

5-Iodo-1,2,3-trimethoxybenzene is used as a primary and secondary intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodo-1,2,3-trimethoxybenzene is used as a primary and secondary intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text