Thermo Scientific Chemicals

5-Methyl-2-hexanone, 99%, Thermo Scientific Chemicals

Catalog number: L17584.AP
500 mL, Each
Thermo Scientific Chemicals

5-Methyl-2-hexanone, 99%, Thermo Scientific Chemicals

Catalog number: L17584.AP
500 mL, Each
Quantity
Catalog number: L17584.AP
also known as L17584-AP
Price (USD)
Price: 48.20
Online price: 40.65
Your price:
Quantity
-

Chemical Identifiers

CAS
110-12-3
IUPAC Name
5-methylhexan-2-one
Molecular Formula
C7H14O
InChI Key
FFWSICBKRCICMR-UHFFFAOYSA-N
SMILES
CC(C)CCC(C)=O
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Refractive Index
1.4060-1.4090 @ 20°C

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (5.4 g/L)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text