Thermo Scientific Chemicals

tert-Butyl isocyanide, 98%, Thermo Scientific Chemicals

Catalog number: L19747.03
1 g, Each
Thermo Scientific Chemicals

tert-Butyl isocyanide, 98%, Thermo Scientific Chemicals

Catalog number: L19747.03
1 g, Each
Quantity
Catalog number: L19747.03
also known as L19747-03
Price (USD)
Price: 52.10
Your price:
Quantity
-

Chemical Identifiers

CAS
7188-38-7
IUPAC Name
2-isocyano-2-methylpropane
Molecular Formula
C5H9N
InChI Key
FAGLEPBREOXSAC-UHFFFAOYSA-N
SMILES
CC(C)(C)[N+]#[C-]
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.3735-1.3775 @ 20?C

Description

tert-Butyl isocyanide is used in a tin-free procedure for alkyl radical reactions in the presence of a free-radical initiator. It is a useful intermediate in multicomponent reactions. It is also used in the synthesis of coumarines, 4H-chromenes, isoxazolines and to trap 2-cyclopropylidene-1,3-diones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl isocyanide is used in a tin-free procedure for alkyl radical reactions in the presence of a free-radical initiator. It is a useful intermediate in multicomponent reactions. It is also used in the synthesis of coumarines, 4H-chromenes, isoxazolines and to trap 2-cyclopropylidene-1,3-diones.

Solubility
Soluble in organic solvents such as ethanol, methanol, ether, toluene and dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text