Thermo Scientific Chemicals

2-Aminopyridine-4-methanol, 97%, Thermo Scientific Chemicals

Catalog number: L19846.03
1 g, Each
Thermo Scientific Chemicals

2-Aminopyridine-4-methanol, 97%, Thermo Scientific Chemicals

Catalog number: L19846.03
1 g, Each
Quantity
Catalog number: L19846.03
also known as L19846-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
105250-17-7
Form
powder
Assay (Silylated GC)
> 96.0%
Appearance (Color)
Pale cream to yellow or brown

Description

2-Aminopyridine-4-methanol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminopyridine-4-methanol is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text