Thermo Scientific Chemicals

1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%, Thermo Scientific Chemicals

Catalog number: L20001.22
100 g, Each
Thermo Scientific Chemicals

1,2-Cyclohexanedicarboxylic anhydride, cis + trans, 97%, Thermo Scientific Chemicals

Catalog number: L20001.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L20001.22
also known as L20001-22
Price (USD)
Price: 33.80
Your price:
Quantity
-

Chemical Identifiers

CAS
85-42-7
IUPAC Name
octahydro-2-benzofuran-1,3-dione
Molecular Formula
C8H10O3
InChI Key
MUTGBJKUEZFXGO-UHFFFAOYNA-N
SMILES
O=C1OC(=O)C2CCCCC12
Form
Lumps or fused solid
Appearance (Color)
Colorless to white
Identification (FTIR)
Conforms (non-U.S. specification)
Assay (GC)
≥96.0%
Melting Point (clear melt)
30.0-39.0°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text