Thermo Scientific Chemicals

3-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L20169.03
1 g, Each
Thermo Scientific Chemicals

3-Cyanobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: L20169.03
1 g, Each
Quantity
Catalog number: L20169.03
also known as L20169-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
56542-67-7
IUPAC Name
3-cyanobenzene-1-sulfonyl chloride
Molecular Formula
C7H4ClNO2S
InChI Key
BHNRGBRMCNHNQD-UHFFFAOYSA-N
SMILES
ClS(=O)(=O)C1=CC=CC(=C1)C#N
Appearance (Color)
White to yellow
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)
45.0-54.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text