Thermo Scientific Chemicals

Methyl 2-amino-5-chlorobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: L20187.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2-amino-5-chlorobenzoate, 98+%, Thermo Scientific Chemicals

Catalog number: L20187.06
5 g, Each
Quantity
Catalog number: L20187.06
also known as L20187-06
Price (USD)
Price: 88.70
Online price: 75.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5202-89-1
IUPAC Name
methyl 2-amino-5-chlorobenzoate
Molecular Formula
C8H8ClNO2
InChI Key
IGHVUURTQGBABT-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(Cl)=CC=C1N
Appearance (Color)
White to cream
Assay (GC)
≥98.0%
Melting Point (clear melt)
67.0-73.0?C
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text