Thermo Scientific Chemicals

Tri-n-butyl(vinyl)tin, 96%, Thermo Scientific Chemicals

Catalog number: L20215.03
1 g, Each
Thermo Scientific Chemicals

Tri-n-butyl(vinyl)tin, 96%, Thermo Scientific Chemicals

Catalog number: L20215.03
1 g, Each
Quantity
Catalog number: L20215.03
also known as L20215-03
Price (USD)

Chemical Identifiers

CAS
7486-35-3
IUPAC Name
tributyl(ethenyl)stannane
Molecular Formula
C14H30Sn
InChI Key
QIWRFOJWQSSRJZ-UHFFFAOYSA-N
SMILES
CCCC[Sn](CCCC)(CCCC)C=C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥95.0%
Form
Liquid
Refractive Index
1.4750-1.4810 @ 20?C

Description

Tributyl(vinyl)tin is used as a reagent in the palladium catalyzed synthesis of allyl and benzyl ethers. It is also a vinyl nucleophile for bromoacetylene and bromoaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tributyl(vinyl)tin is used as a reagent in the palladium catalyzed synthesis of allyl and benzyl ethers. It is also a vinyl nucleophile for bromoacetylene and bromoaromatics.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text