(S)-(-)-3-Butyn-2-ol, 99%
(S)-(-)-3-Butyn-2-ol, 99%
(S)-(-)-3-Butyn-2-ol, 99%
Thermo Scientific Chemicals

(S)-(-)-3-Butyn-2-ol, 99%

CAS: 2914-69-4 | C4H6O | 70.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L20345.03
also known as L20345-03
Price (EUR)
99,30
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
99,30
Each
Specifications
Beilstein4652597
CAS2914-69-4
Chemical Name or Material(S)-(-)-3-Butyn-2-ol
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
Density0.891
View more
(S)-(-)-3-Butyn-2-ol is used to prepare (S)-(-)-2-tert-butyldimethylsiloxybut-3-yne. It finds application in ruthenium-catalyzed polymerization to get helical polyacetylenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Butyn-2-ol is used to prepare (S)-(-)-2-tert-butyldimethylsiloxybut-3-yne. It finds application in ruthenium-catalyzed polymerization to get helical polyacetylenes.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Li, N.; Wang, T. Y.; Gong, L. Z.; Zhang, L. Gold-Catalyzed Multiple Cascade Reaction of 2-Alkynylphenylazides with Propargyl Alcohols. Chem. Eur. J. 2015, 21 (9), 3585-3588.
  2. Pal, I.; Chaudhari, S. R.; Suryaprakash, N. R. Chiral discrimination of secondary alcohols and carboxylic acids by NMR spectroscopy. Magn. Reson. Chem. 2015, 53 (2), 142-146.