A part of the MAPK Pathway, ERK2 is phosphorylated at Thr185 and Tyr187 by the mitogen-activated protein kinase kinases 2 (MAP2K2) . For use in combination with the LanthaScreen™ ERK2 A375 cell line (K1798) to evaluate inhibitors in medium to high throughput screening applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Conjugate
Tb (Terbium)/Lanthanide Chelates
Quantity
1 mg
Excitation/Emission
340/490, 520
Reactivity
Human
Shipping Condition
Dry Ice
Readout
End Point
Reactivity abrv
Hu
Target Gene
Ser/Thr Kinases, Tyrosine Kinases
Product Line
LanthaScreen™
Contents & Storage
Terbium-labeled pERK2 (pThr185/pTyr187) antibody in HEPES buffered saline. Store at -20°C.