Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Remel enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.
Volume (Metric) | 5 x 250 mL |
Type | Gram Iodine |
Quantity | 5/Pk. |
Certifications/Compliance | CE Marked |
Unit Size | Each |
Volume (Metric) | 3.785 L |
Type | Gram Iodine |
Quantity | Each |
Certifications/Compliance | CE Marked |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Type | Quantity | Certifications/Compliance | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R40234 | Each | Gram Iodine | 5/Pk. | CE Marked | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R40235 | Each | Gram Iodine | Each | CE Marked | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Remel enhanced gram stain reagents are balanced to give a distinct gram-positive reaction and are less prone to over decolorization.