Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COX-10 EBGEX Assays ›
Gene Symbol:
cox-10
Gene Name:
Protoheme IX farnesyltransferase, mitochondrial
Gene Aliases:
CELE_Y46G5A.2
Chromosome Location:
Chr.II: 12679954 - 12686766 on Build WBcel235
UniGene ID:
Cel.14063
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Type:
4
Detection Label:
Alexa Fluor 488
ProductType:
ViewRNA Cell Plus Probe Set
Assay ID
VF4-4052387-VCP
Size
M: 440 uL
L: 1.44 mL
Availability
Made To Order
Catalog #
VX-01
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: cox-10
Interrogated Sequence
Translated Protein
IMAGE Clone ID
RefSeq
NM_064307.6
NP_496708.2
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
transferase
Gene Ontology Categories:
Function(s)
Process(es)
heme biosynthetic process
cellular respiration
heme O biosynthetic process
farnesyltranstransferase activity
protoheme IX farnesyltransferase activity
transferase activity
transferase activity, transferring alkyl or aryl (other than methyl) groups
Back To Top
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-whlt2:80/10.239.162.50:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0