Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
IL12B
Transcript:
Assay Name:
Assay ID
CRISPR1013637_LV
Size
200 µL,1x10⁸ TU/mL
200 µL,1x10⁸ TU/mL
Availability
Made To Order
Catalog #
A32042
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
interleukin superfamily
signaling molecule
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of T cell mediated cytotoxicity
positive regulation of defense response to virus by host
natural killer cell activation involved in immune response
positive regulation of T-helper 1 type immune response
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
negative regulation of inflammatory response to antigenic stimulus
cell cycle arrest
response to UV-B
positive regulation of activation of JAK2 kinase activity
cell migration
cytokine-mediated signaling pathway
sensory perception of pain
sexual reproduction
natural killer cell activation
negative regulation of interleukin-10 production
negative regulation of interleukin-17 production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-17 production
positive regulation of tumor necrosis factor production
positive regulation of natural killer cell activation
positive regulation of natural killer cell proliferation
positive regulation of mononuclear cell proliferation
positive regulation of tissue remodeling
positive regulation of smooth muscle cell apoptotic process
regulation of cytokine biosynthetic process
T-helper 1 type immune response
T-helper cell differentiation
interferon-gamma biosynthetic process
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
positive regulation of NF-kappaB import into nucleus
regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat4 protein
positive regulation of tyrosine phosphorylation of Stat5 protein
defense response to protozoan
positive regulation of memory T cell differentiation
negative regulation of growth of symbiont in host
positive regulation of interferon-gamma biosynthetic process
positive regulation of osteoclast differentiation
positive regulation of cell adhesion
negative regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
positive regulation of inflammatory response
defense response to Gram-negative bacterium
positive regulation of NK T cell activation
positive regulation of NK T cell proliferation
defense response to virus
cellular response to lipopolysaccharide
cellular response to interferon-gamma
positive regulation of T-helper 17 type immune response
positive regulation of T-helper 17 cell lineage commitment
cytokine receptor activity
cytokine activity
interleukin-12 receptor binding
protein binding
growth factor activity
interleukin-12 alpha subunit binding
identical protein binding
protein homodimerization activity
interleukin-23 receptor binding
protein heterodimerization activity
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-7dvbj:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0