Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Jak3
Transcript:
Assay Name:
Assay ID
CRISPR466274_SG
Size
3 nmol Synthetic sgRNA
3 nmol Synthetic sgRNA
Availability
Made To Order
Catalog #
A35510
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
negative regulation of dendritic cell cytokine production
protein phosphorylation
enzyme linked receptor protein signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of cytosolic calcium ion concentration
tyrosine phosphorylation of STAT protein
phosphorylation
cell migration
peptidyl-tyrosine phosphorylation
cytokine-mediated signaling pathway
B cell differentiation
erythrocyte differentiation
negative regulation of interleukin-10 production
negative regulation of interleukin-12 production
intracellular signal transduction
interleukin-4-mediated signaling pathway
peptidyl-tyrosine autophosphorylation
positive regulation of T cell proliferation
positive regulation of activated T cell proliferation
tyrosine phosphorylation of Stat5 protein
T cell homeostasis
innate immune response
negative regulation of FasL biosynthetic process
negative regulation of T-helper 1 cell differentiation
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
lymph node development
positive regulation of immune response
negative regulation of T cell activation
positive regulation of calcium ion transport
regulation of T cell apoptotic process
negative regulation of thymocyte apoptotic process
response to interleukin-2
response to interleukin-4
response to interleukin-15
response to interleukin-9
positive regulation of dendritic cell apoptotic process
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein phosphatase binding
hormone receptor binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-86647:80/10.239.162.121:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0