Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Ngfr
Transcript:
Assay Name:
Assay ID
CRISPR550213_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine receptor
receptor
tumor necrosis factor receptor
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
apoptotic process
signal transduction
multicellular organism development
nervous system development
axon guidance
central nervous system development
response to wounding
regulation of gene expression
positive regulation of cell death
negative regulation of neuron projection development
programmed cell death
detection of temperature stimulus
sensory perception of pain
nerve development
cell differentiation
hair follicle morphogenesis
positive regulation of myelination
positive regulation of synaptic transmission, cholinergic
circadian regulation of gene expression
cellular response to oxidative stress
positive regulation of Rho protein signal transduction
negative regulation of fibroblast growth factor receptor signaling pathway
positive regulation of odontogenesis of dentin-containing tooth
glucose homeostasis
positive regulation of apoptotic process
positive regulation of MAPK cascade
negative regulation of neuron apoptotic process
skin development
positive regulation of fibroblast proliferation
rhythmic process
negative regulation of hair follicle development
positive regulation of protein kinase B signaling
negative regulation of mitochondrial depolarization
positive regulation of synaptic transmission, glutamatergic
negative regulation of cell death
positive regulation of neuron death
regulation of reactive oxygen species metabolic process
positive regulation of apoptotic signaling pathway
regulation of glucose import in response to insulin stimulus
beta-amyloid binding
death receptor activity
neurotrophin TRKA receptor binding
protein binding
Rab GTPase binding
ubiquitin protein ligase binding
neurotrophin binding
nerve growth factor binding
preprotein binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0