Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Gene:
Tnf
Transcript:
Assay Name:
Assay ID
CRISPR562713_CR
Size
2 nmol Synthetic crRNA
2 nmol Synthetic crRNA
Availability
Made To Order
Catalog #
A35509
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
signaling molecule
tumor necrosis factor family member
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
activation of MAPKKK activity
activation of MAPK activity
cell activation
positive regulation of cytokine production
regulation of protein phosphorylation
positive regulation of protein phosphorylation
negative regulation of L-glutamate transport
chronic inflammatory response to antigenic stimulus
negative regulation of cytokine secretion involved in immune response
positive regulation of chronic inflammatory response to antigenic stimulus
positive regulation of humoral immune response mediated by circulating immunoglobulin
glucose metabolic process
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
obsolete transformed cell apoptotic process
defense response
inflammatory response
immune response
humoral immune response
signal transduction
JNK cascade
multicellular organism development
cell proliferation
positive regulation of cell proliferation
negative regulation of cell proliferation
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
response to virus
organ morphogenesis
positive regulation of gene expression
negative regulation of gene expression
negative regulation of alkaline phosphatase activity
calcium-mediated signaling
extracellular matrix organization
osteoclast differentiation
sequestering of triglyceride
positive regulation of protein complex assembly
positive regulation of fever generation
lipopolysaccharide-mediated signaling pathway
negative regulation of interleukin-6 production
positive regulation of chemokine production
positive regulation of interferon-gamma production
positive regulation of interleukin-18 production
positive regulation of interleukin-6 production
receptor biosynthetic process
positive regulation of peptidyl-serine phosphorylation
tumor necrosis factor-mediated signaling pathway
positive regulation of heterotypic cell-cell adhesion
regulation of cell proliferation
positive regulation of NF-kappaB import into nucleus
defense response to bacterium
positive regulation of apoptotic process
positive regulation of programmed cell death
regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of protein complex disassembly
positive regulation of protein complex disassembly
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of MAP kinase activity
protein kinase B signaling
positive regulation of JUN kinase activity
positive regulation of neuron apoptotic process
negative regulation of growth of symbiont in host
negative regulation of viral genome replication
positive regulation of chemokine biosynthetic process
cellular extravasation
positive regulation of interleukin-8 biosynthetic process
positive regulation of nitric oxide biosynthetic process
negative regulation of myoblast differentiation
negative regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of osteoclast differentiation
positive regulation of action potential
positive regulation of cell adhesion
positive regulation of mitotic nuclear division
positive regulation of protein kinase activity
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of translational initiation by iron
negative regulation of glucose import
positive regulation of JNK cascade
positive regulation of smooth muscle cell proliferation
regulation of protein secretion
positive regulation of cytokine secretion
positive regulation of inflammatory response
positive regulation of phagocytosis
regulation of insulin secretion
positive regulation of synaptic transmission
defense response to Gram-positive bacterium
leukocyte migration
leukocyte tethering or rolling
detection of mechanical stimulus involved in sensory perception of pain
negative regulation of lipid catabolic process
regulation of immunoglobulin secretion
positive regulation of membrane protein ectodomain proteolysis
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
positive regulation of protein transport
response to glucocorticoid
positive regulation of NFAT protein import into nucleus
positive regulation of hair follicle development
positive regulation of protein kinase B signaling
positive regulation of vitamin D biosynthetic process
positive regulation of calcidiol 1-monooxygenase activity
epithelial cell proliferation involved in salivary gland morphogenesis
regulation of branching involved in salivary gland morphogenesis
negative regulation of branching involved in lung morphogenesis
cellular response to lipopolysaccharide
cellular response to amino acid stimulus
cellular response to nicotine
cellular response to organic cyclic compound
positive regulation of podosome assembly
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
necroptotic signaling pathway
positive regulation of protein localization to cell surface
positive regulation of chemokine (C-X-C motif) ligand 2 production
regulation of reactive oxygen species metabolic process
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protease binding
cytokine activity
tumor necrosis factor receptor binding
protein binding
identical protein binding
transcription regulatory region DNA binding
Back To Top
Related Products
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0